Search
Search
| Gene Symbol: USP33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678223.3 | XP_005678280.2 | 7 - 8 | 604 | 68 | |
| XM_005678224.3 | XP_005678281.2 | 7 - 8 | 604 | 68 | ||
| XM_005678225.3 | XP_005678282.2 | 7 - 8 | 837 | 68 | ||
| XM_018045640.1 | XP_017901129.1 | 7 - 8 | 776 | 68 | ||
| GenBank mRNA | JT687133.1 | - | 6 - 7 | 969 | 68 | |
Set Membership: |
|