Search
Search
| Gene Symbol: PANX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013975740.2 | XP_013831194.2 | 1 - 2 | 585 | 74 | |
| GenBank mRNA | JO439359.2 | - | 1 - 2 | 353 | 74 | |
| JO609572.2 | - | 1 - 2 | 364 | 74 | ||
Set Membership: |
|