Search
Search
| Gene Symbol: OAZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285559.1 | NP_001272488.1 | 3 - 4 | 616 | 118 | |
| GenBank mRNA | EF564268.1 | - | 2 - 3 | 199 | 118 | |
| JO579793.1 | - | 3 - 4 | 616 | 118 | ||
| JO634402.2 | - | 3 - 4 | 550 | 118 | ||
| KA318171.1 | - | 2 - 3 | 651 | 118 | ||
Set Membership: |
|