Search
Search

ST3GAL1| Gene Symbol: ST3GAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058592.1 | XP_017914081.1 | 4 - 5 | 1397 | 70 | |
| XM_018058593.1 | XP_017914082.1 | 3 - 4 | 1342 | 70 | ||
| XM_018058594.1 | XP_017914083.1 | 3 - 4 | 1245 | 70 | ||
| XM_018058595.1 | XP_017914084.1 | 2 - 3 | 1190 | 70 | ||
| XM_018058596.1 | XP_017914085.1 | 4 - 5 | 1398 | 70 | ||
| GenBank mRNA | JO428052.1 | - | 7 - 8 | 2245 | 70 | |
| JT682916.1 | - | 4 - 5 | 1303 | 70 | ||
Set Membership: |
|