Search
Search

SDCCAG8| Gene Symbol: SDCCAG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690561.3 | XP_005690618.2 | 2 - 3 | 462 | 88 | |
| XM_013970100.2 | XP_013825554.2 | 2 - 3 | 463 | 88 | ||
| XM_013970101.2 | XP_013825555.2 | 2 - 3 | 463 | 88 | ||
| XM_013970102.2 | XP_013825556.2 | 2 - 3 | 462 | 88 | ||
| XM_013970103.2 | XP_013825557.2 | 2 - 3 | 464 | 88 | ||
| XM_018060257.1 | XP_017915746.1 | 2 - 3 | 464 | 88 | ||
| XM_018060258.1 | XP_017915747.1 | 2 - 3 | 464 | 88 | ||
Set Membership: |
|