Search
Search
| Gene Symbol: PDAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040209.1 | XP_017895698.1 | 3 - 4 | 333 | 92 | |
| GenBank mRNA | JO580117.1 | - | 3 - 4 | 330 | 92 | |
| JR130719.1 | - | 3909 | 92 | |||
Set Membership: |
|