Search
Search

ARHGAP45| Gene Symbol: ARHGAP45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050476.1 | XP_017905965.1 | 21 - 22 | 3876 | 58 | |
| XM_018050477.1 | XP_017905966.1 | 21 - 22 | 3650 | 58 | ||
| XM_018050478.1 | XP_017905967.1 | 20 - 21 | 2700 | 58 | ||
| XM_018050479.1 | XP_017905968.1 | 21 - 22 | 3356 | 58 | ||
| GenBank mRNA | JO581934.1 | - | 21 - 22 | 3187 | 58 | |
| JO581935.1 | - | 11 - 12 | 2855 | 58 | ||
Set Membership: |
|