Search
Search

PIK3C3| Gene Symbol: PIK3C3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696968.3 | XP_005697025.1 | 12 - 13 | 1507 | 100 | |
| XM_005696969.3 | XP_005697026.1 | 11 - 12 | 1318 | 100 | ||
| XM_013974301.2 | XP_013829755.1 | 12 - 13 | 1414 | 100 | ||
| GenBank mRNA | JO585165.2 | - | 13 - 14 | 1806 | 100 | |
| JO585166.2 | - | 13 - 14 | 1806 | 100 | ||
Set Membership: |
|