Search
Search

ADAMTS2| Gene Symbol: ADAMTS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051595.1 | XP_017907084.1 | 6 - 7 | 1242 | 65 | |
| XM_018051596.1 | XP_017907085.1 | 6 - 7 | 1245 | 65 | ||
Set Membership: |
|