Search
Search

TCEANC2| Gene Symbol: TCEANC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678376.3 | XP_005678433.2 | 3 - 4 | 437 | 112 | |
| XM_005678377.3 | XP_005678434.2 | 3 - 4 | 3303 | 112 | ||
Set Membership: |
|