Search
Search
| Gene Symbol: FCGRT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062789.1 | XP_017918278.1 | 6 - 7 | 1125 | 82 | |
| XM_018062790.1 | XP_017918279.1 | 5 - 6 | 1197 | 82 | ||
| GenBank mRNA | JO579294.1 | - | 6 - 7 | 1182 | 82 | |
Set Membership: |
|