Search
Search

SNAPC4| Gene Symbol: SNAPC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056095.1 | XP_017911584.1 | 11 - 12 | 1191 | 54 | |
| XM_018056096.1 | XP_017911585.1 | 11 - 12 | 1653 | 54 | ||
| XM_018056097.1 | XP_017911586.1 | 10 - 11 | 2103 | 54 | ||
| XM_018056098.1 | XP_017911587.1 | 9 - 10 | 1010 | 54 | ||
| GenBank mRNA | JO576291.1 | - | 4 - 5 | 490 | 54 | |
Set Membership: |
|