Search
Search

DNAJA3| Gene Symbol: DNAJA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697929.3 | XP_005697986.3 | 6 - 7 | 964 | 74 | |
| XM_018040843.1 | XP_017896332.1 | 6 - 7 | 963 | 74 | ||
| GenBank mRNA | JO551359.2 | - | 6 - 7 | 893 | 74 | |
Set Membership: |
|