Search
Search

LOC102186442| Gene Symbol: LOC102186442 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039319.1 | XP_017894808.1 | 4 - 5 | 787 | 91 | |
| GenBank mRNA | JO589527.2 | - | 1 - 2 | 394 | 91 | |
Set Membership: |
|