Search
Search
| Gene Symbol: RUSC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052123.1 | XP_017907612.1 | 3 - 4 | 3086 | 64 | |
| XM_018052125.1 | XP_017907614.1 | 3 - 4 | 3302 | 64 | ||
| XM_018052126.1 | XP_017907615.1 | 3 - 4 | 2904 | 64 | ||
| XM_018052127.1 | XP_017907616.1 | 4 - 5 | 3372 | 64 | ||
| XM_018052128.1 | XP_017907617.1 | 3 - 4 | 3464 | 64 | ||
| XM_018052129.1 | XP_017907618.1 | 3 - 4 | 3053 | 64 | ||
| XM_018052130.1 | XP_017907619.1 | 3 - 4 | 3268 | 64 | ||
| GenBank mRNA | JO645306.2 | - | 1 - 2 | 872 | 64 | |
| JO645307.2 | - | 3 - 4 | 3023 | 64 | ||
| JT630860.1 | - | 3 - 4 | 2907 | 64 | ||
Set Membership: |
|