Search
Search
| Gene Symbol: SUGCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679346.3 | XP_005679403.1 | 3 - 4 | 377 | 95 | |
| XM_013963404.2 | XP_013818858.1 | 3 - 4 | 377 | 95 | ||
| XM_018046996.1 | XP_017902485.1 | 3 - 4 | 377 | 95 | ||
Set Membership: |
|