Search
Search

RAB11FIP4| Gene Symbol: RAB11FIP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064266.1 | XP_017919755.1 | 11 - 12 | 1546 | 63 | |
| XM_018064267.1 | XP_017919756.1 | 9 - 10 | 1332 | 63 | ||
Set Membership: |
|