Search
Search
| Gene Symbol: IQCE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040500.1 | XP_017895989.1 | 12 - 13 | 1110 | 65 | |
| XM_018040501.1 | XP_017895990.1 | 12 - 13 | 1095 | 65 | ||
| XM_018040503.1 | XP_017895992.1 | 12 - 13 | 1110 | 65 | ||
| XM_018040504.1 | XP_017895993.1 | 11 - 12 | 975 | 65 | ||
| XM_018040505.1 | XP_017895994.1 | 10 - 11 | 945 | 65 | ||
| XM_018040506.1 | XP_017895995.1 | 10 - 11 | 950 | 65 | ||
| XM_018040507.1 | XP_017895996.1 | 13 - 14 | 1381 | 65 | ||
| XM_018040508.1 | XP_017895997.1 | 9 - 10 | 810 | 65 | ||
Set Membership: |
|