Search
Search
| Gene Symbol: RBM25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686044.3 | XP_005686101.1 | 14 - 15 | 1919 | 58 | |
| GenBank mRNA | JO427382.1 | - | 5 - 6 | 869 | 58 | |
| JT707200.1 | - | 16 - 17 | 2369 | 58 | ||
Set Membership: |
|