Search
Search
| Gene Symbol: MARK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695427.3 | XP_005695484.2 | 1 - 2 | 681 | 56 | |
| XM_005695428.3 | XP_005695485.2 | 1 - 2 | 235 | 56 | ||
| XM_005695429.3 | XP_005695486.2 | 1 - 2 | 681 | 56 | ||
| XM_005695430.2 | XP_005695487.2 | 1 - 2 | 681 | 56 | ||
| XM_005695431.3 | XP_005695488.2 | 1 - 2 | 681 | 56 | ||
| XM_013973238.2 | XP_013828692.2 | 1 - 2 | 235 | 56 | ||
| XM_013973240.2 | XP_013828694.2 | 1 - 2 | 234 | 56 | ||
| XM_018066071.1 | XP_017921560.1 | 1 - 2 | 239 | 56 | ||
| XM_018066072.1 | XP_017921561.1 | 1 - 2 | 238 | 56 | ||
| XM_018066073.1 | XP_017921562.1 | 1 - 2 | 239 | 56 | ||
| XM_018066074.1 | XP_017921563.1 | 1 - 2 | 240 | 56 | ||
| XM_018066075.1 | XP_017921564.1 | 1 - 2 | 238 | 56 | ||
| XM_018066076.1 | XP_017921565.1 | 1 - 2 | 235 | 56 | ||
Set Membership: |
|