Search
Search
| Gene Symbol: ZFPM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061841.1 | XP_017917330.1 | 3 - 4 | 353 | 89 | |
| XM_018061842.1 | XP_017917331.1 | 3 - 4 | 353 | 89 | ||
| XM_018061843.1 | XP_017917332.1 | 3 - 4 | 353 | 89 | ||
| XM_018061844.1 | XP_017917333.1 | 3 - 4 | 353 | 89 | ||
| XM_018061845.1 | XP_017917334.1 | 3 - 4 | 305 | 89 | ||
Set Membership: |
|