Search
Search

ARHGAP28| Gene Symbol: ARHGAP28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039589.1 | XP_017895078.1 | 10 - 11 | 1624 | 67 | |
| XM_018039590.1 | XP_017895079.1 | 10 - 11 | 1624 | 67 | ||
| XM_018039591.1 | XP_017895080.1 | 9 - 10 | 1531 | 67 | ||
| XM_018039592.1 | XP_017895081.1 | 9 - 10 | 1531 | 67 | ||
| XM_018039593.1 | XP_017895082.1 | 10 - 11 | 1739 | 67 | ||
Set Membership: |
|