Search
Search

ZNF667| Gene Symbol: ZNF667 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063429.1 | XP_017918918.1 | 4 - 5 | 495 | 65 | |
| XM_018063430.1 | XP_017918919.1 | 4 - 5 | 594 | 65 | ||
| GenBank mRNA | KA317939.1 | - | 2 - 3 | 490 | 65 | |
Set Membership: |
|