Search
Search

EDRF1| Gene Symbol: EDRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756697.2 | XP_002756743.1 | 7 - 8 | 1203 | 98 | |
| XM_002756698.2 | XP_002756744.1 | 8 - 9 | 1305 | 98 | ||
Set Membership: |
|