Search
Search

SLC22A7| Gene Symbol: SLC22A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746569.3 | XP_002746615.1 | 4 - 5 | 924 | 67 | |
| XM_008994461.2 | XP_008992709.1 | 4 - 5 | 926 | 67 | ||
| XM_008994462.2 | XP_008992710.1 | 4 - 5 | 935 | 67 | ||
| XM_008994463.2 | XP_008992711.1 | 4 - 5 | 936 | 67 | ||
| XM_008994464.2 | XP_008992712.1 | 4 - 5 | 937 | 67 | ||
| XM_008994465.2 | XP_008992713.1 | 5 - 6 | 699 | 67 | ||
| XM_008994466.2 | XP_008992714.1 | 5 - 6 | 688 | 67 | ||
Set Membership: |
|