Search
Search

KCNK10| Gene Symbol: KCNK10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754176.4 | XP_002754222.1 | 4 - 5 | 1149 | 87 | |
| XM_009006424.2 | XP_009004672.1 | 4 - 5 | 1400 | 87 | ||
Set Membership: |
|