Search
Search

UBE2R2| Gene Symbol: UBE2R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743021.3 | XP_002743067.1 | 4 - 5 | 1070 | 93 | |
| XM_017974156.1 | XP_017829645.1 | 4 - 5 | 429 | 93 | ||
Set Membership: |
|