Search
Search

NOP14| Gene Symbol: NOP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008993686.2 | XP_008991934.1 | 5 - 6 | 942 | 60 | |
| XM_008993687.2 | XP_008991935.1 | 5 - 6 | 942 | 60 | ||
| XM_008993688.2 | XP_008991936.1 | 5 - 6 | 942 | 60 | ||
| XM_008993689.2 | XP_008991937.1 | 5 - 6 | 942 | 60 | ||
Set Membership: |
|