Search
Search

pot| Gene Symbol: pot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258706.2 | NP_001245635.1 | 2 - 3 | 429 | 109 | |
| NM_132540.3 | NP_572768.1 | 2 - 3 | 429 | 109 | ||
| NM_167313.3 | NP_727583.1 | 3 - 4 | 464 | 109 | ||
| GenBank mRNA | AY862156.1 | - | 2 - 3 | 405 | 109 | |
| BT010304.1 | - | 2 - 3 | 402 | 109 | ||
Set Membership: |
|