Search
Search

cdc123| Gene Symbol: cdc123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200721.1 | NP_957015.1 | 1 - 2 | 87 | 131 | |
| XM_009299951.1 | XP_009298226.1 | 1 - 2 | 122 | 131 | ||
| GenBank mRNA | BC059483.1 | - | 1 - 2 | 87 | 131 | |
| BC071332.1 | - | 1 - 2 | 100 | 131 | ||
| BC152266.1 | - | 1 - 2 | 98 | 131 | ||
Set Membership: |
|