Search
Search

tonsl| Gene Symbol: tonsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111148.1 | NP_001104618.1 | 24 - 25 | 3924 | 76 | |
| GenBank mRNA | BC155547.1 | - | 24 - 25 | 3924 | 76 | |
Set Membership: |
|