Search
Search

smad5| Gene Symbol: smad5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001346309.1 | NP_001333238.1 | 3 - 4 | 1177 | 69 | |
| NM_131368.3 | NP_571443.3 | 4 - 5 | 1262 | 69 | ||
| XM_005173164.4 | XP_005173221.1 | 4 - 5 | 1256 | 69 | ||
| GenBank mRNA | AF127920.1 | - | 2 - 3 | 1032 | 69 | |
| AF168775.1 | - | 2 - 3 | 667 | 69 | ||
| BC050481.1 | - | 4 - 5 | 1252 | 69 | ||
| BC065644.1 | - | 4 - 5 | 1144 | 69 | ||
| BC154311.1 | - | 2 - 3 | 761 | 69 | ||
| BC155776.1 | - | 3 - 4 | 1144 | 69 | ||
| BC165695.1 | - | 2 - 3 | 701 | 69 | ||
| DT073830.1 | - | 2 - 3 | 314 | 69 | ||
Set Membership: |
|