Search
Search

CAMKMT| Gene Symbol: CAMKMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001917869.5 | XP_001917904.3 | 812 | 87 | ||
| XM_023619712.1 | XP_023475480.1 | 821 | 87 | |||
| XM_023619713.1 | XP_023475481.1 | 819 | 87 | |||
| XM_023619714.1 | XP_023475482.1 | 817 | 87 | |||
| XM_023619715.1 | XP_023475483.1 | 632 | 87 | |||
| GenBank mRNA | JL636749.1 | - | 7 - 8 | 444 | 87 | |
Set Membership: |
|