Search
Search

SH3GLB1| Gene Symbol: SH3GLB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023641722.1 | XP_023497490.1 | 564 | 138 | ||
| XM_023641723.1 | XP_023497491.1 | 563 | 138 | |||
| XM_023641724.1 | XP_023497492.1 | 564 | 138 | |||
| XM_023641725.1 | XP_023497493.1 | 562 | 138 | |||
| XM_023641726.1 | XP_023497494.1 | 563 | 138 | |||
| XM_023641727.1 | XP_023497495.1 | 562 | 138 | |||
| XM_023641728.1 | XP_023497496.1 | 561 | 138 | |||
Set Membership: |
|