Search
Search
| Gene Symbol: PBX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023628884.1 | XP_023484652.1 | 1299 | 102 | ||
| XM_023628885.1 | XP_023484653.1 | 1184 | 102 | |||
| XM_023628886.1 | XP_023484654.1 | 3159 | 102 | |||
| GenBank mRNA | JL618790.1 | - | 5 - 6 | 733 | 102 | |
Set Membership: |
|