Search
Search
| Gene Symbol: TFRC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081913.1 | NP_001075382.1 | 10 - 11 | 1332 | 67 | |
| XM_005601764.3 | XP_005601821.1 | 1501 | 67 | |||
| XM_023623131.1 | XP_023478899.1 | 1433 | 67 | |||
| GenBank mRNA | DQ284764.1 | - | 10 - 11 | 1332 | 67 | |
| JL627344.1 | - | 2 - 3 | 148 | 67 | ||
Set Membership: |
|