Search
Search

ITGAV| Gene Symbol: ITGAV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023622166.1 | XP_023477934.1 | 125 | |||
| XM_023622167.1 | XP_023477935.1 | 125 | ||||
| GenBank mRNA | AB266840.1 | - | 1 - 2 | 127 | 125 | |
Set Membership: |
|