Search
Search

ITGAL| Gene Symbol: ITGAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005598775.3 | XP_005598832.1 | 1990 | 69 | ||
| XM_014730016.2 | XP_014585502.1 | 1990 | 69 | |||
| GenBank mRNA | EU881921.1 | - | 2 - 3 | 198 | 69 | |
Set Membership: |
|