Search
Search
| Gene Symbol: POT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005609347.3 | XP_005609404.1 | 1836 | 72 | ||
| XM_023639598.1 | XP_023495366.1 | 2130 | 72 | |||
| XM_023639599.1 | XP_023495367.1 | 2083 | 72 | |||
| XM_023639600.1 | XP_023495368.1 | 2127 | 72 | |||
| XM_023639601.1 | XP_023495369.1 | 2080 | 72 | |||
| XM_023639602.1 | XP_023495370.1 | 1965 | 72 | |||
| XM_023639604.1 | XP_023495372.1 | 1905 | 72 | |||
| XM_023639605.1 | XP_023495373.1 | 1721 | 72 | |||
| XM_023639606.1 | XP_023495374.1 | 2135 | 72 | |||
| XM_023639607.1 | XP_023495375.1 | 1825 | 72 | |||
| XM_023639608.1 | XP_023495376.1 | 1756 | 72 | |||
| GenBank mRNA | JL639327.1 | - | 1 - 2 | 142 | 72 | |
Set Membership: |
|