Search
Search
| Gene Symbol: GGCX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001498891.6 | XP_001498941.2 | 1272 | 91 | ||
| XM_005599862.3 | XP_005599919.1 | 1207 | 91 | |||
| XM_023618787.1 | XP_023474555.1 | 992 | 91 | |||
| XM_023618788.1 | XP_023474556.1 | 1272 | 91 | |||
Set Membership: |
|