Search
Search

DNAH6| Gene Symbol: DNAH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001916886.4 | XP_001916921.2 | 11744 | 93 | ||
| XM_023618816.1 | XP_023474584.1 | 11711 | 93 | |||
| XM_023618817.1 | XP_023474585.1 | 11723 | 93 | |||