Search
Search

CIDEC| Gene Symbol: CIDEC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005600382.3 | XP_005600439.1 | 479 | 105 | ||
| XM_023619999.1 | XP_023475767.1 | 1247 | 105 | |||
| XM_023620000.1 | XP_023475768.1 | 1246 | 105 | |||
| XM_023620001.1 | XP_023475769.1 | 576 | 105 | |||
Set Membership: |
|