Search
Search

GOLGA4| Gene Symbol: GOLGA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001489013.5 | XP_001489063.1 | 1936 | 119 | ||
| XM_014731787.2 | XP_014587273.1 | 2119 | 119 | |||
| XM_014731788.2 | XP_014587274.1 | 2119 | 119 | |||
| XM_014731789.2 | XP_014587275.1 | 2035 | 119 | |||
| XM_014731791.2 | XP_014587277.1 | 2002 | 119 | |||
| XM_014731792.2 | XP_014587278.1 | 2002 | 119 | |||
Set Membership: |
|