Search
Search

IQSEC3| Gene Symbol: IQSEC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023642969.1 | XP_023498737.1 | 2633 | 109 | ||
| XM_023642970.1 | XP_023498738.1 | 2633 | 109 | |||
| XM_023642971.1 | XP_023498739.1 | 2633 | 109 | |||
| XM_023642972.1 | XP_023498740.1 | 2633 | 109 | |||
| XM_023642973.1 | XP_023498741.1 | 2633 | 109 | |||
Set Membership: |
|