Search
Search
| Gene Symbol: TANK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001493298.5 | XP_001493348.1 | 813 | 89 | ||
| XM_005601539.3 | XP_005601596.1 | 670 | 89 | |||
| XM_005601541.3 | XP_005601598.1 | 667 | 89 | |||
| XM_005601542.3 | XP_005601599.1 | 439 | 89 | |||
| XM_023623003.1 | XP_023478771.1 | 685 | 89 | |||
| XM_023623004.1 | XP_023478772.1 | 831 | 89 | |||
| XM_023623005.1 | XP_023478773.1 | 682 | 89 | |||
Set Membership: |
|