Search
Search
| Gene Symbol: POLQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014732866.2 | XP_014588352.2 | 311 | 76 | ||
| XM_023623672.1 | XP_023479440.1 | 311 | 76 | |||
| XM_023623675.1 | XP_023479443.1 | 311 | 76 | |||
Set Membership: |
|