Search
Search

KIAA0319L| Gene Symbol: KIAA0319L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503679.6 | XP_001503729.1 | 2217 | 88 | ||
| XM_005607204.3 | XP_005607261.1 | 2294 | 88 | |||
| XM_005607206.3 | XP_005607263.1 | 2128 | 88 | |||
| XM_005607207.3 | XP_005607264.1 | 2051 | 88 | |||
| XM_014737290.2 | XP_014592776.1 | 2123 | 88 | |||
| XM_023633557.1 | XP_023489325.1 | 2030 | 88 | |||
| XM_023633562.1 | XP_023489330.1 | 1891 | 88 | |||