Search
Search
| Gene Symbol: FRS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001501157.5 | XP_001501207.1 | 784 | 98 | ||
| XM_023624916.1 | XP_023480684.1 | 1969 | 98 | |||
| XM_023624917.1 | XP_023480685.1 | 734 | 98 | |||
| XM_023624918.1 | XP_023480686.1 | 874 | 98 | |||
| XM_023624920.1 | XP_023480688.1 | 973 | 98 | |||
| XM_023624921.1 | XP_023480689.1 | 622 | 98 | |||
| XM_023624922.1 | XP_023480690.1 | 799 | 98 | |||