Search
Search

RTEL1| Gene Symbol: RTEL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023626938.1 | XP_023482706.1 | 1428 | 65 | ||
| XM_023626939.1 | XP_023482707.1 | 1428 | 65 | |||
| XM_023626940.1 | XP_023482708.1 | 1425 | 65 | |||
| XM_023626941.1 | XP_023482709.1 | 1424 | 65 | |||
| XM_023626942.1 | XP_023482710.1 | 1428 | 65 | |||
| XM_023626943.1 | XP_023482711.1 | 1428 | 65 | |||
| XM_023626944.1 | XP_023482712.1 | 1428 | 65 | |||
| XM_023626945.1 | XP_023482713.1 | 1430 | 65 | |||
| XM_023626946.1 | XP_023482714.1 | 1428 | 65 | |||
| XM_023626947.1 | XP_023482715.1 | 1431 | 65 | |||
| XM_023626948.1 | XP_023482716.1 | 1430 | 65 | |||
Set Membership: |
|