Search
Search
| Gene Symbol: MED6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001487916.6 | XP_001487966.1 | 351 | 75 | ||
| XM_005605281.3 | XP_005605338.1 | 357 | 75 | |||
| XM_014735676.2 | XP_014591162.1 | 357 | 75 | |||
| XM_023628011.1 | XP_023483779.1 | 355 | 75 | |||
| GenBank mRNA | JL615890.1 | - | 2 - 3 | 230 | 75 | |
Set Membership: |
|